siRNA Details
Gene Symbol: Sly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201530.2 | NP_963288.2 | 5 | 296 | ||
GenBank mRNA | BC049626.1 | - | 5 | 296 | 6705850 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201530.2 | NP_963288.2 | 5 | 296 | ||
GenBank mRNA | BC049626.1 | - | 5 | 296 | 6705850 |