siRNA Details
| Gene Symbol: Taar8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010837.1 | NP_001010837.1 | 1 | 577 | ||
| GenBank mRNA | BC139105.1 | - | 1 | 603 | 8862127 | |
| BC139106.1 | - | 1 | 603 | 8862128 | ||
More Information
Other Species Targeted
Set Membership: |
|

Taar8b
Genomic Map