siRNA Details
| Gene Symbol: Poc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027740.6 | NP_082016.1 | 4 | 404 | ||
| GenBank mRNA | AK016991.1 | - | 3 | 328 | ||
| AK030180.1 | - | 3 | 298 | |||
| AK045302.1 | - | 4 | 383 | |||
| AK050233.1 | - | 4 | 473 | |||
| AK090075.1 | - | 4 | 407 | |||
| BC050888.1 | - | 3 | 402 | |||
| BC099926.1 | - | 4 | 491 | |||
| BC144819.1 | - | 4 | 493 | |||
| BC145801.1 | - | 4 | 493 | 40131104 | ||

Poc1b
Genomic Map