siRNA Details
| Gene Symbol: Atxn7l3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033474.2 | NP_001028646.2 | 1 | 1167 | ||
| GenBank mRNA | AK014824.1 | - | 1 | 1063 | ||
| BC047210.1 | - | 1 | 1138 | |||
| BC058561.1 | - | 1 | 1099 | |||
| BC079579.1 | - | 1 | 1178 | |||
| BC094390.1 | - | 1 | 1126 | |||

Atxn7l3b
Genomic Map