siRNA Details
| Gene Symbol: RARRES3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004585.4 | NP_004576.2 | 3, 4 | 453 | ||
| GenBank mRNA | AB030815.1 | - | 3, 4 | 409 | ||
| AB453252.1 | - | 2, 3 | 385 | |||
| AF060228.1 | - | 3, 4 | 414 | |||
| AF092922.1 | - | 3, 4 | 446 | |||
| AK312110.1 | - | 3, 4 | 438 | |||
| BC009678.1 | - | 3, 4 | 433 | 3877277 | ||

RARRES3
Genomic Map