siRNA Details
Gene Symbol: Ankfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080933.1 | NP_001074402.1 | 2 | 192 | ||
GenBank mRNA | AK190172.1 | - | 2 | 153 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ankfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080933.1 | NP_001074402.1 | 2 | 192 | ||
GenBank mRNA | AK190172.1 | - | 2 | 153 |