siRNA Details
| Gene Symbol: Olfr128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206816.1 | NP_996552.1 | 1 | 807 | ||
| GenBank mRNA | BC139087.1 | - | 1 | 837 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr128| Gene Symbol: Olfr128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206816.1 | NP_996552.1 | 1 | 807 | ||
| GenBank mRNA | BC139087.1 | - | 1 | 837 | ||