siRNA Details
| Gene Symbol: Serpinb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198680.2 | NP_941373.1 | 6 | 617 | ||
| GenBank mRNA | AK003220.1 | - | 6 | 618 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinb3b| Gene Symbol: Serpinb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198680.2 | NP_941373.1 | 6 | 617 | ||
| GenBank mRNA | AK003220.1 | - | 6 | 618 | ||