siRNA Details
| Gene Symbol: Ankrd63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081971.1 | NP_001075440.1 | 1 | 2419 | ||
| GenBank mRNA | AK087787.1 | - | 1 | 1106 | ||
| AK135101.1 | - | 1 | 2320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd63| Gene Symbol: Ankrd63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081971.1 | NP_001075440.1 | 1 | 2419 | ||
| GenBank mRNA | AK087787.1 | - | 1 | 1106 | ||
| AK135101.1 | - | 1 | 2320 | |||