siRNA Details
| Gene Symbol: Gm5283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC172000.1 | - | 1 | 177 | 9054286 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5283| Gene Symbol: Gm5283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC172000.1 | - | 1 | 177 | 9054286 | |