siRNA Details
| Gene Symbol: Glipr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027450.3 | NP_081726.1 | 4 | 307 | ||
| GenBank mRNA | AK017557.1 | - | 4 | 284 | ||
| AK089960.1 | - | 4 | 287 | |||
| AK141459.1 | - | 4 | 296 | |||
| AK154433.1 | - | 4 | 285 | |||
| AK167595.1 | - | 4 | 307 | |||
| BC031750.1 | - | 4 | 307 | 4037002 | ||

Glipr2
Genomic Map