siRNA Details
| Gene Symbol: Fndc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027402.3 | NP_081678.1 | 5 | 589 | ||
| GenBank mRNA | AJ401376.1 | - | 5 | 589 | ||
| AK005096.1 | - | 5 | 651 | |||
| BC109183.1 | - | 5 | 585 | 40050064 | ||
| BC109184.1 | - | 5 | 589 | 40050065 | ||

Fndc5
Genomic Map