siRNA Details
| Gene Symbol: RBBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135255.1 | NP_001128727.1 | 7 | 929 | ||
| NM_001135256.1 | NP_001128728.1 | 7 | 832 | |||
| NM_005610.2 | NP_005601.1 | 7 | 932 | |||
| GenBank mRNA | AK222779.1 | - | 7 | 835 | ||
| AK299251.1 | - | 7 | 832 | |||
| AK299410.1 | - | 5 | 543 | |||
| AK312571.1 | - | 7 | 870 | |||
| BC003092.1 | - | 7 | 859 | 3356558 | ||
| BC015123.1 | - | 5 | 593 | |||
| BC053904.1 | - | 7 | 845 | 6526200 | ||
| BC075836.1 | - | 7 | 800 | 6338602 | ||
| BT007309.1 | - | 7 | 773 | |||
| X71810.1 | - | 7 | 846 | |||
| X74262.1 | - | 7 | 857 | |||

RBBP4
Genomic Map