siRNA Details
| Gene Symbol: Trim56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201373.4 | NP_958761.1 | 3 | 1909 | ||
| GenBank mRNA | AK037350.1 | - | 3 | 1910 | ||
| BC045615.1 | - | 3 | 1899 | 4976515 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim56| Gene Symbol: Trim56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201373.4 | NP_958761.1 | 3 | 1909 | ||
| GenBank mRNA | AK037350.1 | - | 3 | 1910 | ||
| BC045615.1 | - | 3 | 1899 | 4976515 | ||