siRNA Details
| Gene Symbol: Ccdc155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201374.2 | NP_958762.2 | 11, 12 | 1326 | ||
| GenBank mRNA | AB725603.1 | - | 11, 12 | 1137 | ||
| BC050196.1 | - | 11, 12 | 1326 | 30008752 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc155| Gene Symbol: Ccdc155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201374.2 | NP_958762.2 | 11, 12 | 1326 | ||
| GenBank mRNA | AB725603.1 | - | 11, 12 | 1137 | ||
| BC050196.1 | - | 11, 12 | 1326 | 30008752 | ||