siRNA Details
Gene Symbol: Gm4133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167158.1 | NP_001160630.1 | 1 | 463 |
Gene Symbol: Gm4187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167159.1 | NP_001160631.1 | 1 | 463 |
Gene Symbol: Vmn1r119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166708.1 | NP_001160180.1 | 1 | 463 |
