siRNA Details
| Gene Symbol: Pglyrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165968.1 | NP_001159440.1 | 5 | 595 | ||
| NM_207263.2 | NP_997146.2 | 5 | 595 | |||
| GenBank mRNA | AY518699.1 | - | 5 | 549 | ||
| BC120840.1 | - | 5 | 590 | 40129763 | ||
| BC144902.1 | - | 5 | 595 | 9053450 | ||
| BC144927.1 | - | 4 | 476 | |||

Pglyrp4
Genomic Map