siRNA Details
| Gene Symbol: Tnpo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U21453.1 | - | 23 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnpo3| Gene Symbol: Tnpo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U21453.1 | - | 23 | |||