siRNA Details
| Gene Symbol: BC061237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198677.1 | NP_941079.1 | 3 | 262 | ||
| GenBank mRNA | BC061237.1 | - | 3 | 262 | 6705671 | |
| Gene Symbol: 1700001F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027940.2 | NP_082216.1 | 3 | 247 | ||
| GenBank mRNA | AK005587.1 | - | 3 | 248 | ||
| BC061060.1 | - | 3 | 229 | 6770724 | ||

BC061237
Genomic Map