siRNA Details
| Gene Symbol: Gm5382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034100.1 | NP_001029272.1 | 1 | 304 | ||
| GenBank mRNA | BC100482.1 | - | 1 | 304 | 30918325 | |
| Gene Symbol: Gm14474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242947.1 | NP_001229876.1 | 1 | 300 | ||

Gm5382
Genomic Map