siRNA Details
| Gene Symbol: Tex28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126488.2 | NP_001119960.1 | 3 | 574 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex28| Gene Symbol: Tex28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126488.2 | NP_001119960.1 | 3 | 574 | ||