siRNA Details
| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028964.4 | NP_083240.2 | 8 | 968 | ||
| GenBank mRNA | CD805796.1 | - | 3 | 435 | ||
| CJ070275.1 | - | 2 | 212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx29| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028964.4 | NP_083240.2 | 8 | 968 | ||
| GenBank mRNA | CD805796.1 | - | 3 | 435 | ||
| CJ070275.1 | - | 2 | 212 | |||