siRNA Details
Gene Symbol: Cdsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008424.2 | NP_001008424.2 | 2 | 1662 | ||
GenBank mRNA | AK133029.1 | - | 1 | 930 | ||
BC055373.1 | - | 2 | 1659 | 30058959 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008424.2 | NP_001008424.2 | 2 | 1662 | ||
GenBank mRNA | AK133029.1 | - | 1 | 930 | ||
BC055373.1 | - | 2 | 1659 | 30058959 |