siRNA Details
| Gene Symbol: Thoc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008425.1 | NP_001008425.1 | 11 | 985 | ||
| GenBank mRNA | AK089746.1 | - | 8 | 1313 | ||
| BC085131.1 | - | 11 | 985 | 5715435 | ||
| BC089551.1 | - | 12 | 985 | 30303151 | ||

Thoc6
Genomic Map