siRNA Details
| Gene Symbol: Tmtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198967.5 | NP_945318.2 | 6 | 1117 | ||
| GenBank mRNA | AK046396.1 | - | 6 | 1118 | ||
| AK137545.1 | - | 6 | 1089 | |||
| AK162239.1 | - | 6 | 773 | |||
| BC145513.1 | - | 4 | 448 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmtc1| Gene Symbol: Tmtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198967.5 | NP_945318.2 | 6 | 1117 | ||
| GenBank mRNA | AK046396.1 | - | 6 | 1118 | ||
| AK137545.1 | - | 6 | 1089 | |||
| AK162239.1 | - | 6 | 773 | |||
| BC145513.1 | - | 4 | 448 | |||