siRNA Details
| Gene Symbol: Tmtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198967.5 | NP_945318.2 | 12 | 1938 | ||
| GenBank mRNA | AK046396.1 | - | 11 | 1825 | ||
| AK137545.1 | - | 12 | 1910 | |||
| AK162239.1 | - | 12 | 1594 | |||
| BC023818.1 | - | 1 | 99 | |||
| BC145513.1 | - | 10 | 1269 | |||

Tmtc1
Genomic Map