siRNA Details
| Gene Symbol: Tas2r104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207011.1 | NP_996894.1 | 1 | 188 | ||
| GenBank mRNA | BC148229.1 | - | 1 | 192 | 40142081 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r104| Gene Symbol: Tas2r104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207011.1 | NP_996894.1 | 1 | 188 | ||
| GenBank mRNA | BC148229.1 | - | 1 | 192 | 40142081 | |