siRNA Details
| Gene Symbol: Tas2r106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207016.1 | NP_996899.1 | 1 | 583 | ||
| GenBank mRNA | BC107179.1 | - | 1 | 582 | ||
| BC107180.1 | - | 1 | 583 | 40050680 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r106| Gene Symbol: Tas2r106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207016.1 | NP_996899.1 | 1 | 583 | ||
| GenBank mRNA | BC107179.1 | - | 1 | 582 | ||
| BC107180.1 | - | 1 | 583 | 40050680 | ||