siRNA Details
| Gene Symbol: Tas2r109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207017.1 | NP_996900.1 | 1 | 208 | ||
| GenBank mRNA | BC127604.1 | - | 1 | 205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r109| Gene Symbol: Tas2r109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207017.1 | NP_996900.1 | 1 | 208 | ||
| GenBank mRNA | BC127604.1 | - | 1 | 205 | ||