siRNA Details
| Gene Symbol: Tas2r118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207022.1 | NP_996905.1 | 1 | 54 | ||
| GenBank mRNA | BC104400.2 | - | 1 | 46 | ||
| BC104401.1 | - | 1 | 54 | 40050472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r118| Gene Symbol: Tas2r118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207022.1 | NP_996905.1 | 1 | 54 | ||
| GenBank mRNA | BC104400.2 | - | 1 | 46 | ||
| BC104401.1 | - | 1 | 54 | 40050472 | ||