siRNA Details
| Gene Symbol: Tas2r121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207024.1 | NP_996907.1 | 1 | 791 | ||
| GenBank mRNA | BC104410.1 | - | 1 | 791 | 40050579 | |
| BC104411.1 | - | 1 | 791 | 40050580 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r121| Gene Symbol: Tas2r121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207024.1 | NP_996907.1 | 1 | 791 | ||
| GenBank mRNA | BC104410.1 | - | 1 | 791 | 40050579 | |
| BC104411.1 | - | 1 | 791 | 40050580 | ||