siRNA Details
| Gene Symbol: Tas2r131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207030.1 | NP_996913.1 | 1 | 655 | ||
| GenBank mRNA | BC145798.1 | - | 1 | 730 | 40131101 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r131| Gene Symbol: Tas2r131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207030.1 | NP_996913.1 | 1 | 655 | ||
| GenBank mRNA | BC145798.1 | - | 1 | 730 | 40131101 | |