siRNA Details
| Gene Symbol: Ifnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199157.2 | NP_954608.1 | 1 | 322 | ||
| GenBank mRNA | BC099376.1 | - | 1 | 322 | 30713338 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifnk| Gene Symbol: Ifnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199157.2 | NP_954608.1 | 1 | 322 | ||
| GenBank mRNA | BC099376.1 | - | 1 | 322 | 30713338 | |