siRNA Details
| Gene Symbol: Tas2r143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001452.1 | NP_001001452.1 | 1 | 766 | ||
| GenBank mRNA | BC111103.1 | - | 1 | 765 | ||
| BC148239.1 | - | 1 | 766 | 40050382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r143| Gene Symbol: Tas2r143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001452.1 | NP_001001452.1 | 1 | 766 | ||
| GenBank mRNA | BC111103.1 | - | 1 | 765 | ||
| BC148239.1 | - | 1 | 766 | 40050382 | ||