siRNA Details
| Gene Symbol: Cd300c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199225.1 | NP_954695.1 | 2 | 140 | ||
| GenBank mRNA | AY457052.1 | - | 2 | 140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd300c| Gene Symbol: Cd300c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199225.1 | NP_954695.1 | 2 | 140 | ||
| GenBank mRNA | AY457052.1 | - | 2 | 140 | ||