siRNA Details
| Gene Symbol: Ugt1a7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201642.4 | NP_964004.1 | 1 | 892 | ||
| GenBank mRNA | AK128918.1 | - | 1 | 818 | ||
| AK144599.1 | - | 1 | 784 | |||
| AK153157.1 | - | 1 | 782 | |||
| AK154973.1 | - | 1 | 815 | |||
| AK159496.1 | - | 1 | 830 | |||
| AK162317.1 | - | 1 | 816 | |||
| AK165465.1 | - | 1 | 816 | |||
| BC141205.1 | - | 1 | 801 | 9055817 | ||
| BC141206.1 | - | 1 | 801 | 9055818 | ||

Ugt1a7c
Genomic Map