siRNA Details
| Gene Symbol: Ugt1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201645.2 | NP_964007.2 | 1 | 143 | ||
| GenBank mRNA | AY227194.1 | - | 1 | 143 | ||
| BC093516.1 | - | 1 | 105 | 5135493 | ||
| L02333.1 | - | 1 | 115 | |||
| S64760.1 | - | 1 | 115 | |||

Ugt1a1
Genomic Map