siRNA Details
| Gene Symbol: Flrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201518.4 | NP_958926.1 | 2 | 1908 | ||
| GenBank mRNA | AK041311.1 | - | 2 | 1881 | ||
| AK076530.1 | - | 1 | 1348 | |||
| AK135149.1 | - | 2 | 1909 | |||
| AK136973.1 | - | 2 | 1902 | |||
| AK140831.1 | - | 2 | 2141 | |||
| AK172945.1 | - | 1 | 1068 | |||
| AY495669.1 | - | 1 | 1347 | |||
| BC067058.1 | - | 2 | 1889 | |||
| BC096471.1 | - | 2 | 1876 | 3582155 | ||
| BC138297.1 | - | 1 | 1356 | 8861316 | ||
| BC138298.1 | - | 1 | 1356 | 8861317 | ||

Flrt2
Genomic Map