siRNA Details
| Gene Symbol: Tmsb15b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080967.4 | NP_001074436.1 | 3 | 486 | ||
| Gene Symbol: Tmsb15l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207267.4 | NP_997150.1 | 4 | 626 | ||

Tmsb15b2
Genomic Map