siRNA Details
| Gene Symbol: Tmsb15l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207267.4 | NP_997150.1 | 2, 3 | 217 | ||
| GenBank mRNA | AK029707.1 | - | 2, 3 | 194 | ||
| BB623661.1 | - | 2, 3 | 213 | |||
| BC096054.1 | - | 2, 3 | 150 | 3373135 | ||
| BE625563.1 | - | 2, 3 | 142 | |||

Tmsb15l
Genomic Map