siRNA Details
Gene Symbol: Tdpoz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207271.2 | NP_997154.2 | 1 | 323 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tdpoz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207271.2 | NP_997154.2 | 1 | 323 |