siRNA Details
| Gene Symbol: Tdpoz4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207272.2 | NP_997155.2 | 1 | 329 | ||
| GenBank mRNA | BC115626.1 | - | 1 | 329 | 40102845 | |
| BC115627.1 | - | 1 | 325 | |||
| Gene Symbol: Gm4858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177694.1 | NP_001171165.1 | 1 | 329 | ||
| GenBank mRNA | BC172161.1 | - | 1 | 313 | ||

Tdpoz4
Genomic Map