siRNA Details
| Gene Symbol: Plcxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281812.1 | NP_001268741.1 | 4 | 385 | ||
| NM_207279.2 | NP_997162.2 | 5 | 552 | |||
| GenBank mRNA | AK039459.1 | - | 3 | 292 | ||
| AK149435.1 | - | 4 | 386 | |||
| BC034116.1 | - | 2 | 313 | |||
| BC039627.1 | - | 5 | 552 | 5038680 | ||

Plcxd1
Genomic Map