siRNA Details
| Gene Symbol: Agr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207531.3 | NP_997414.2 | 4 | 346 | ||
| GenBank mRNA | AK053177.1 | - | 4 | 275 | ||
| BC023499.1 | - | 4 | 347 | 5008294 | ||
| BE650904.1 | - | 4 | 346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agr3| Gene Symbol: Agr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207531.3 | NP_997414.2 | 4 | 346 | ||
| GenBank mRNA | AK053177.1 | - | 4 | 275 | ||
| BC023499.1 | - | 4 | 347 | 5008294 | ||
| BE650904.1 | - | 4 | 346 | |||