siRNA Details
| Gene Symbol: Clec3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007223.3 | NP_001007224.1 | 3 | 377 | ||
| GenBank mRNA | BC147139.1 | - | 3 | 377 | 9056169 | |
| BC147140.1 | - | 3 | 377 | 9056166 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec3a| Gene Symbol: Clec3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007223.3 | NP_001007224.1 | 3 | 377 | ||
| GenBank mRNA | BC147139.1 | - | 3 | 377 | 9056169 | |
| BC147140.1 | - | 3 | 377 | 9056166 | ||