siRNA Details
| Gene Symbol: Vmn1r179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207545.1 | NP_997428.1 | 1 | 838 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r179| Gene Symbol: Vmn1r179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207545.1 | NP_997428.1 | 1 | 838 | ||