siRNA Details
| Gene Symbol: RBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323517.1 | NP_001310446.1 | 3, 4 | 411 | ||
| NM_001323518.1 | NP_001310447.1 | 3, 4 | 263 | |||
| NM_006744.3 | NP_006735.2 | 3, 4 | 325 | |||
| GenBank mRNA | BC020633.1 | - | 3, 4 | 325 | 4767481 | |
| BG565176.1 | - | 3, 4 | 264 | |||
| X00129.1 | - | 3, 4 | 286 | |||

RBP4
Genomic Map