siRNA Details
| Gene Symbol: Olfr100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207673.1 | NP_997556.1 | 1 | 55 | ||
| GenBank mRNA | BC111097.1 | - | 1 | 54 | ||
| BC127233.1 | - | 1 | 55 | 40054913 | ||
| BC127234.1 | - | 1 | 55 | 40054914 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr100| Gene Symbol: Olfr100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207673.1 | NP_997556.1 | 1 | 55 | ||
| GenBank mRNA | BC111097.1 | - | 1 | 54 | ||
| BC127233.1 | - | 1 | 55 | 40054913 | ||
| BC127234.1 | - | 1 | 55 | 40054914 | ||