siRNA Details
| Gene Symbol: Olfr100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207673.1 | NP_997556.1 | 1 | 809 | ||
| GenBank mRNA | BC111097.1 | - | 1 | 808 | ||
| BC127233.1 | - | 1 | 809 | 40054913 | ||
| BC127234.1 | - | 1 | 809 | 40054914 | ||
| Gene Symbol: Olfr102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011721.2 | NP_001011721.2 | 1 | 864 | ||
| GenBank mRNA | BC139114.1 | - | 1 | 864 | 8862136 | |
| BC139115.1 | - | 1 | 864 | 8862137 | ||

Olfr100
Genomic Map