siRNA Details
Gene Symbol: Olfr118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213721.2 | NP_998886.2 | 1 | 278 | ||
GenBank mRNA | BC128012.1 | - | 1 | 282 | 40136025 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213721.2 | NP_998886.2 | 1 | 278 | ||
GenBank mRNA | BC128012.1 | - | 1 | 282 | 40136025 |